Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGACTGCTTGGGCACGCCAGCAGC[A/G]TTCATGTAGCTGTGATGGGAGGGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601408 | ||||||||||||||||||||
Literature Links: |
KAT6A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KAT6A - lysine acetyltransferase 6A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305878.1 | 6620 | Intron | NP_001292807.1 | |||
NM_006766.4 | 6620 | Silent Mutation | AAC,AAT | N,N 1988 | NP_006757.2 | |
XM_011544656.2 | 6620 | Silent Mutation | AAC,AAT | N,N 2032 | XP_011542958.1 | |
XM_011544657.2 | 6620 | Silent Mutation | AAC,AAT | N,N 2032 | XP_011542959.1 | |
XM_011544658.2 | 6620 | Silent Mutation | AAC,AAT | N,N 2032 | XP_011542960.1 | |
XM_011544659.2 | 6620 | Silent Mutation | AAC,AAT | N,N 2025 | XP_011542961.1 | |
XM_017013863.1 | 6620 | Silent Mutation | AAC,AAT | N,N 1988 | XP_016869352.1 | |
XM_017013864.1 | 6620 | Silent Mutation | AAC,AAT | N,N 1988 | XP_016869353.1 |