Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCACCACCTCCGAGTACTTGAC[A/G]GGGGTTCCAGGACCGGGGATCCGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 107272 MIM: 157660 MIM: 604964 | ||||||||||||||||||||
Literature Links: |
ARHGEF39 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARHGEF39 - Rho guanine nucleotide exchange factor 39 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CCDC107 - coiled-coil domain containing 107 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CD72 - CD72 molecule | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101926948 - uncharacterized LOC101926948 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RMRP - RNA component of mitochondrial RNA processing endoribonuclease | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIT1 - signaling threshold regulating transmembrane adaptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014450.2 | 532 | Silent Mutation | CCC,CCT | P,P 145 | NP_055265.1 |