Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAAGCTGGAAATATTAATATAACC[C/T]CTGGTAAGATGCAAATTGTGAGCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614533 MIM: 607795 | ||||||||||||||||||||
Literature Links: |
CDC26 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDC26 - cell division cycle 26 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRPF4 - pre-mRNA processing factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244926.1 | 315 | Missense Mutation | CCT,TCT | P,S 68 | NP_001231855.1 | |
NM_001322266.1 | 315 | UTR 5 | NP_001309195.1 | |||
NM_001322267.1 | 315 | UTR 5 | NP_001309196.1 | |||
NM_004697.4 | 315 | Missense Mutation | CCT,TCT | P,S 69 | NP_004688.2 |