Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGGGGGCCGCCGTCCCCGTGGA[A/G]CTGAGGCGGGAGCGACGCATGGTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604890 | ||||||||||||||||||||
Literature Links: |
GTF3C5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GTF3C5 - general transcription factor IIIC subunit 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122823.1 | 388 | Silent Mutation | GAA,GAG | E,E 17 | NP_001116295.1 | |
NM_001286709.1 | 388 | UTR 5 | NP_001273638.1 | |||
NM_012087.3 | 388 | Silent Mutation | GAA,GAG | E,E 17 | NP_036219.2 | |
XM_005272234.3 | 388 | Silent Mutation | GAA,GAG | E,E 17 | XP_005272291.1 | |
XM_011519201.1 | 388 | Intron | XP_011517503.1 | |||
XM_017015315.1 | 388 | Intron | XP_016870804.1 | |||
XM_017015316.1 | 388 | Intron | XP_016870805.1 |
LOC105376306 - uncharacterized LOC105376306 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |