Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGTGTTGGCGCCACCCTCGATGG[C/T]GATGCCCAGGGTGGCCGCACTTTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605729 MIM: 607928 | ||||||||||||||||||||
Literature Links: |
AKNA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AKNA - AT-hook transcription factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WHRN - whirlin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083885.2 | 3226 | Missense Mutation | ACC,GCC | T,A 446 | NP_001077354.2 | |
NM_001173425.1 | 3226 | Missense Mutation | ACC,GCC | T,A 828 | NP_001166896.1 | |
NM_015404.3 | 3226 | Missense Mutation | ACC,GCC | T,A 829 | NP_056219.3 | |
XM_011518484.1 | 3226 | Missense Mutation | ACC,GCC | T,A 840 | XP_011516786.1 | |
XM_011518485.1 | 3226 | Missense Mutation | ACC,GCC | T,A 840 | XP_011516787.1 | |
XM_011518486.1 | 3226 | Missense Mutation | ACC,GCC | T,A 839 | XP_011516788.1 | |
XM_011518487.1 | 3226 | Missense Mutation | ACC,GCC | T,A 798 | XP_011516789.1 | |
XM_011518488.1 | 3226 | Missense Mutation | ACC,GCC | T,A 759 | XP_011516790.1 | |
XM_011518489.2 | 3226 | Intron | XP_011516791.1 | |||
XM_011518491.2 | 3226 | Intron | XP_011516793.1 | |||
XM_011518492.1 | 3226 | Intron | XP_011516794.1 | |||
XM_011518494.2 | 3226 | Intron | XP_011516796.1 | |||
XM_011518495.1 | 3226 | Missense Mutation | ACC,GCC | T,A 399 | XP_011516797.1 |