Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCAGGCGGAACAGCACAACGTTC[C/T]CGAGCCGCCACGGCCTGCTGCTGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602745 | ||||||||||||||||||||
Literature Links: |
FAM122A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM122A - family with sequence similarity 122A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138333.4 | 355 | Missense Mutation | CCG,TCG | P,S 108 | NP_612206.4 |
PIP5K1B - phosphatidylinositol-4-phosphate 5-kinase type 1 beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278253.1 | 355 | Intron | NP_001265182.1 | |||
NM_003558.3 | 355 | Intron | NP_003549.1 | |||
XM_005252261.3 | 355 | Intron | XP_005252318.1 | |||
XM_005252262.4 | 355 | Intron | XP_005252319.1 | |||
XM_006717300.2 | 355 | Intron | XP_006717363.1 | |||
XM_006717301.1 | 355 | Intron | XP_006717364.1 | |||
XM_011519082.2 | 355 | Intron | XP_011517384.1 | |||
XM_011519083.2 | 355 | Intron | XP_011517385.1 | |||
XM_011519084.2 | 355 | Intron | XP_011517386.1 | |||
XM_017015188.1 | 355 | Intron | XP_016870677.1 | |||
XM_017015189.1 | 355 | Intron | XP_016870678.1 | |||
XM_017015190.1 | 355 | Intron | XP_016870679.1 | |||
XM_017015191.1 | 355 | Intron | XP_016870680.1 |