Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCCAAAGCCCTTGTGGCAGTACA[C/G]TTGGGACAGGTAGGTGAGGGTCCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TMEM246 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEM246 - transmembrane protein 246 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303107.1 | 1410 | Missense Mutation | CTG,GTG | L,V 354 | NP_001290036.1 | |
NM_001303108.1 | 1410 | Missense Mutation | CTG,GTG | L,V 354 | NP_001290037.1 | |
NM_032342.2 | 1410 | Missense Mutation | CTG,GTG | L,V 354 | NP_115718.1 | |
XM_017015206.1 | 1410 | Missense Mutation | CTG,GTG | L,V 354 | XP_016870695.1 |
TMEM246-AS1 - TMEM246 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |