Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCCGCAAACATGCTCTTCCCTTC[A/G]GAACGGGATTCTGCTAGGAGCAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 185620 MIM: 185630 MIM: 185660 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SURF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SURF1 - surfeit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SURF2 - surfeit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SURF4 - surfeit 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001280788.1 | 605 | Silent Mutation | TCC,TCT | S,S 121 | NP_001267717.1 | |
NM_001280789.1 | 605 | Silent Mutation | TCC,TCT | S,S 136 | NP_001267718.1 | |
NM_001280790.1 | 605 | Silent Mutation | TCC,TCT | S,S 93 | NP_001267719.1 | |
NM_001280791.1 | 605 | Silent Mutation | TCC,TCT | S,S 93 | NP_001267720.1 | |
NM_001280792.1 | 605 | Intron | NP_001267721.1 | |||
NM_001280793.1 | 605 | Silent Mutation | TCC,TCT | S,S 19 | NP_001267722.1 | |
NM_033161.3 | 605 | Silent Mutation | TCC,TCT | S,S 136 | NP_149351.1 | |
XM_011518943.1 | 605 | Silent Mutation | TCC,TCT | S,S 127 | XP_011517245.1 | |
XM_011518944.2 | 605 | Silent Mutation | TCC,TCT | S,S 127 | XP_011517246.1 | |
XM_011518945.2 | 605 | Intron | XP_011517247.1 | |||
XM_017015062.1 | 605 | Intron | XP_016870551.1 |