Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTGAGAGTCCCCACCACACGCTC[A/G]TTCAGCTGGTCACCACGGGGGTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601895 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR4479 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR4479 - microRNA 4479 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRAF2 - TNF receptor associated factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021138.3 | 616 | Intron | NP_066961.2 | |||
XM_011518973.2 | 616 | Intron | XP_011517275.1 | |||
XM_011518974.2 | 616 | UTR 5 | XP_011517276.1 | |||
XM_011518975.2 | 616 | Intron | XP_011517277.1 | |||
XM_011518976.2 | 616 | Intron | XP_011517278.1 | |||
XM_011518977.2 | 616 | Intron | XP_011517279.1 | |||
XM_011518978.2 | 616 | Intron | XP_011517280.1 | |||
XM_017015094.1 | 616 | Intron | XP_016870583.1 | |||
XM_017015095.1 | 616 | Intron | XP_016870584.1 |