Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCCTTTCCCCGCAGCTGTGAGGT[A/G]GAGCAGTGGGACTCGGATGAGCCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604296 | ||||||||||||||||||||
Literature Links: |
GRHPR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GRHPR - glyoxylate reductase/hydroxypyruvate reductase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012203.1 | 148 | Silent Mutation | GTA,GTG | V,V 31 | NP_036335.1 | |
XM_005251631.1 | 148 | Intron | XP_005251688.1 | |||
XM_011518073.2 | 148 | UTR 5 | XP_011516375.1 | |||
XM_017015320.1 | 148 | Silent Mutation | GTA,GTG | V,V 31 | XP_016870809.1 | |
XM_017015321.1 | 148 | Silent Mutation | GTA,GTG | V,V 31 | XP_016870810.1 | |
XM_017015322.1 | 148 | Intron | XP_016870811.1 | |||
XM_017015323.1 | 148 | UTR 5 | XP_016870812.1 |