Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCGTTCGAGCCCATCACCGTGGG[C/T]CTAGCCATCGGGGCCGCGTCGGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605204 MIM: 608050 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TOR1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TOR1A - torsin family 1 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TOR1B - torsin family 1 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317893.1 | 158 | Silent Mutation | GGC,GGT | G,G 31 | NP_001304822.1 | |
NM_001317894.1 | 158 | Silent Mutation | GGC,GGT | G,G 31 | NP_001304823.1 | |
NM_014506.2 | 158 | Silent Mutation | GGC,GGT | G,G 31 | NP_055321.1 | |
XM_005251927.3 | 158 | Silent Mutation | GGC,GGT | G,G 31 | XP_005251984.1 |