Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATAGAAACTTACAGCTGCTACTGT[A/G]AGCATGTCAAAAACTTTGGTACAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612725 | ||||||||||||||||||||
Literature Links: |
PPP6C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPP6C - protein phosphatase 6 catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001123355.1 | 465 | Silent Mutation | CTC,CTT | L,L 186 | NP_001116827.1 | |
NM_001123369.1 | 465 | Silent Mutation | CTC,CTT | L,L 127 | NP_001116841.1 | |
NM_002721.4 | 465 | Silent Mutation | CTC,CTT | L,L 149 | NP_002712.1 | |
XM_011518847.2 | 465 | Silent Mutation | CTC,CTT | L,L 137 | XP_011517149.1 |