Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGCCAAGTCCTAAGTCCCGCTCTC[C/T]TCAGGCTGAATCTATCGGTCGATCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613492 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACER2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACER2 - alkaline ceramidase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010887.2 | Intron | NP_001010887.2 | ||||
XM_005251448.3 | Intron | XP_005251505.1 | ||||
XM_011517858.1 | Intron | XP_011516160.1 | ||||
XM_011517859.2 | Intron | XP_011516161.1 | ||||
XM_017014694.1 | Intron | XP_016870183.1 |