Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGCGGCGCTGCAAGGGCAGCCT[C/G]CGCCCCAACGGCATCATCGTCATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615051 MIM: 613560 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASB6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASB6 - ankyrin repeat and SOCS box containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202403.1 | 694 | UTR 3 | NP_001189332.1 | |||
NM_017873.3 | 694 | UTR 3 | NP_060343.1 | |||
NM_177999.2 | 694 | UTR 3 | NP_821066.1 |
NTMT1 - N-terminal Xaa-Pro-Lys N-methyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286796.1 | 694 | Silent Mutation | CTC,CTG | L,L 157 | NP_001273725.1 | |
NM_001286797.1 | 694 | Silent Mutation | CTC,CTG | L,L 157 | NP_001273726.1 | |
NM_001286798.1 | 694 | Silent Mutation | CTC,CTG | L,L 157 | NP_001273727.1 | |
NM_001286799.1 | 694 | Silent Mutation | CTC,CTG | L,L 157 | NP_001273728.1 | |
NM_001286800.1 | 694 | Missense Mutation | TCC,TGC | S,C 129 | NP_001273729.1 | |
NM_001286801.1 | 694 | Missense Mutation | TCC,TGC | S,C 129 | NP_001273730.1 | |
NM_001286802.1 | 694 | Silent Mutation | CTC,CTG | L,L 69 | NP_001273731.1 | |
NM_001286803.1 | 694 | Silent Mutation | CTC,CTG | L,L 69 | NP_001273732.1 | |
NM_014064.3 | 694 | Silent Mutation | CTC,CTG | L,L 157 | NP_054783.2 | |
XM_005251939.3 | 694 | Silent Mutation | CTC,CTG | L,L 69 | XP_005251996.1 | |
XM_017014642.1 | 694 | Intron | XP_016870131.1 |