Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAGGGAGATGGAGGAAGGTTTCT[A/G]TATGGGGTGGCTGGCTGGATGAGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TRMO PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRMO - tRNA methyltransferase O | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016481.4 | Intron | NP_057565.3 | ||||
XM_005252047.3 | Intron | XP_005252104.1 | ||||
XM_005252048.4 | Intron | XP_005252105.1 | ||||
XM_005252049.4 | Intron | XP_005252106.1 | ||||
XM_005252050.4 | Intron | XP_005252107.1 | ||||
XM_011518776.2 | Intron | XP_011517078.1 | ||||
XM_011518777.1 | Intron | XP_011517079.1 |