Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACTGCTTAGTAGCCGGAGAACAC[A/G]GGACCGCAAGGCACTGGCTGAGGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610714 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PKN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PKN3 - protein kinase N3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317926.1 | 1112 | Missense Mutation | CAG,CGG | Q,R 210 | NP_001304855.1 | |
NM_013355.4 | 1112 | Missense Mutation | CAG,CGG | Q,R 210 | NP_037487.2 | |
XM_005251946.3 | 1112 | Missense Mutation | CAG,CGG | Q,R 207 | XP_005252003.1 | |
XM_006717080.2 | 1112 | Missense Mutation | CAG,CGG | Q,R 217 | XP_006717143.1 | |
XM_017014649.1 | 1112 | Missense Mutation | CAG,CGG | Q,R 217 | XP_016870138.1 | |
XM_017014650.1 | 1112 | Missense Mutation | CAG,CGG | Q,R 84 | XP_016870139.1 |