Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTCAGTGTTATTCGTGACGGAAG[A/G]CGCCATGTTTGCCATCTCAGGTCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602502 | ||||||||||||||||||||
Literature Links: |
ARPC5L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARPC5L - actin related protein 2/3 complex subunit 5 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GOLGA1 - golgin A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002077.3 | 2344 | Missense Mutation | CCT,TCT | P,S 679 | NP_002068.1 | |
XM_005251929.3 | 2344 | Missense Mutation | CCT,TCT | P,S 679 | XP_005251986.1 | |
XM_006717062.3 | 2344 | Missense Mutation | CCT,TCT | P,S 679 | XP_006717125.1 | |
XM_006717063.3 | 2344 | Missense Mutation | CCT,TCT | P,S 679 | XP_006717126.1 |