Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACCTTCGAGTTTGACGACGGCCA[A/G]TGCGCCCCCAGGTACAGTCTGCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608167 MIM: 610224 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNT1 - potassium sodium-activated channel subfamily T member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001272003.1 | 1551 | Silent Mutation | CAA,CAG | Q,Q 33 | NP_001258932.1 | |
NM_020822.2 | 1551 | Silent Mutation | CAA,CAG | Q,Q 33 | NP_065873.2 | |
XM_011518877.2 | 1551 | UTR 5 | XP_011517179.1 | |||
XM_011518878.2 | 1551 | UTR 5 | XP_011517180.1 | |||
XM_011518879.2 | 1551 | UTR 5 | XP_011517181.1 | |||
XM_011518880.1 | 1551 | Intron | XP_011517182.1 | |||
XM_011518881.2 | 1551 | Intron | XP_011517183.1 | |||
XM_017014931.1 | 1551 | Intron | XP_016870420.1 | |||
XM_017014932.1 | 1551 | Intron | XP_016870421.1 | |||
XM_017014933.1 | 1551 | Intron | XP_016870422.1 |
SOHLH1 - spermatogenesis and oogenesis specific basic helix-loop-helix 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012415.2 | 1551 | Intron | NP_001012415.2 | |||
NM_001101677.1 | 1551 | Intron | NP_001095147.1 |