Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGGCGTCCTGCCTAGAGCTGGC[A/G]CTGGAGGGCGAGCGTCTGTGCAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609491 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DKFZP434A062 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DKFZP434A062 - uncharacterized LOC26102 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPSM1 - G-protein signaling modulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145638.2 | 316 | Silent Mutation | GCA,GCG | A,A 32 | NP_001139110.2 | |
NM_001145639.1 | 316 | Intron | NP_001139111.1 | |||
NM_001200003.1 | 316 | Intron | NP_001186932.1 | |||
NM_015597.5 | 316 | Silent Mutation | GCA,GCG | A,A 32 | NP_056412.5 | |
XM_011518498.2 | 316 | Silent Mutation | GCA,GCG | A,A 64 | XP_011516800.1 | |
XM_017014598.1 | 316 | Silent Mutation | GCA,GCG | A,A 9 | XP_016870087.1 | |
XM_017014599.1 | 316 | Intron | XP_016870088.1 |