Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGGCAAAGCCAAGTACATCCTC[C/T]CTACCGAAACAATATATGTTGGGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603359 | ||||||||||||||||||||
Literature Links: |
MORN5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MORN5 - MORN repeat containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286828.1 | 137 | Missense Mutation | CCT,TCT | P,S 26 | NP_001273757.1 | |
NM_198469.3 | 137 | Missense Mutation | CCT,TCT | P,S 26 | NP_940871.2 | |
XM_005251877.4 | 137 | Missense Mutation | CCT,TCT | P,S 26 | XP_005251934.1 | |
XM_005251878.3 | 137 | Missense Mutation | CCT,TCT | P,S 15 | XP_005251935.1 | |
XM_006717034.3 | 137 | Missense Mutation | CCT,TCT | P,S 26 | XP_006717097.1 | |
XM_011518457.2 | 137 | Missense Mutation | CCT,TCT | P,S 26 | XP_011516759.1 | |
XM_011518458.1 | 137 | Missense Mutation | CCT,TCT | P,S 11 | XP_011516760.1 | |
XM_017014578.1 | 137 | Intron | XP_016870067.1 |
NDUFA8 - NADH:ubiquinone oxidoreductase subunit A8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |