Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCTTCGTAAAGGTGGACACACTA[A/G]GATAGGTTCCTTTTTCTCTTCTACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602938 MIM: 611854 MIM: 603132 | ||||||||||||||||||||
Literature Links: |
BAAT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BAAT - bile acid-CoA:amino acid N-acyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL50 - mitochondrial ribosomal protein L50 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019051.2 | Intron | NP_061924.1 |
ZNF189 - zinc finger protein 189 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |