Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGGTGTTGGACTCCCTGTAACGA[A/G]GCGCCAGGCTGGGGAAGCTGCGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616315 MIM: 606533 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C9orf142 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C9orf142 - chromosome 9 open reading frame 142 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CLIC3 - chloride intracellular channel 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004669.2 | 521 | Missense Mutation | CCT,CTT | P,L 96 | NP_004660.2 | |
XM_017015281.1 | 521 | Missense Mutation | CCT,CTT | P,L 96 | XP_016870770.1 | |
XM_017015282.1 | 521 | Missense Mutation | CCT,CTT | P,L 67 | XP_016870771.1 |
LCNL1 - lipocalin like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |