Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCCCTCTGCACTGCTGTCCGAGG[A/G]GCAGCGTCCATCCAGCTCAGCTGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609740 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHF19 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PHF19 - PHD finger protein 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009936.2 | 1619 | Intron | NP_001009936.1 | |||
NM_001286840.1 | 1619 | Missense Mutation | CCC,TCC | P,S 518 | NP_001273769.1 | |
NM_001286842.1 | 1619 | Missense Mutation | CCC,TCC | P,S 290 | NP_001273771.1 | |
NM_001286843.1 | 1619 | Intron | NP_001273772.1 | |||
NM_015651.2 | 1619 | Missense Mutation | CCC,TCC | P,S 499 | NP_056466.1 | |
XM_005251906.2 | 1619 | Missense Mutation | CCC,TCC | P,S 499 | XP_005251963.1 | |
XM_011518509.2 | 1619 | Missense Mutation | CCC,TCC | P,S 499 | XP_011516811.1 | |
XM_011518511.2 | 1619 | UTR 3 | XP_011516813.1 | |||
XM_011518515.2 | 1619 | Intron | XP_011516817.1 | |||
XM_011518516.2 | 1619 | Intron | XP_011516818.1 | |||
XM_017014612.1 | 1619 | Missense Mutation | CCC,TCC | P,S 499 | XP_016870101.1 | |
XM_017014613.1 | 1619 | Intron | XP_016870102.1 |
PSMD5-AS1 - PSMD5 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |