Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCGGGTAGTGGCACAGGGTGTAGT[C/T]AAGCAGGGCGGCGTTGGCCTTCTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 184757 | ||||||||||||||||||||
Literature Links: |
ADGRD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADGRD2 - adhesion G protein-coupled receptor D2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NR5A1 - nuclear receptor subfamily 5 group A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004959.4 | 1422 | Missense Mutation | AAC,GAC | N,D 403 | NP_004950.2 | |
XM_005251871.3 | 1422 | Missense Mutation | AAC,GAC | N,D 403 | XP_005251928.1 | |
XM_011518455.2 | 1422 | Missense Mutation | AAC,GAC | N,D 403 | XP_011516757.1 |