Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCTCGTACTATTAGGACTAGGAG[C/G]AATTCTGCCATTGTGTGTGCTGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300086 | ||||||||||||||||||||
Literature Links: |
LPAR4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LPAR4 - lysophosphatidic acid receptor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278000.1 | 864 | Missense Mutation | AGC,AGG | S,R 153 | NP_001264929.1 | |
NM_005296.2 | 864 | Missense Mutation | AGC,AGG | S,R 153 | NP_005287.1 | |
XM_005262126.3 | 864 | Missense Mutation | AGC,AGG | S,R 153 | XP_005262183.1 | |
XM_006724639.3 | 864 | Missense Mutation | AGC,AGG | S,R 153 | XP_006724702.1 | |
XM_017029437.1 | 864 | Missense Mutation | AGC,AGG | S,R 153 | XP_016884926.1 | |
XM_017029438.1 | 864 | Missense Mutation | AGC,AGG | S,R 153 | XP_016884927.1 |