Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCCTGCCACTGGGTGTTGGATGC[C/T]ACCTTCAGTGATGGCAGTCTAGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300469 | ||||||||||||||||||||
Literature Links: |
MAGEC3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAGEC3 - MAGE family member C3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138702.1 | 33 | Silent Mutation | GCC,GCT | A,A 11 | NP_619647.1 | |
NM_177456.2 | 33 | Intron | NP_803251.1 | |||
XM_011531267.2 | 33 | Intron | XP_011529569.1 | |||
XM_017029265.1 | 33 | Intron | XP_016884754.1 |