Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGTTGACCTTGTTACAGGACTGG[A/T]GTCGGGGGGAACACCTGAACACCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300675 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PNMA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PNMA3 - paraneoplastic Ma antigen 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282535.1 | 364 | Missense Mutation | AGT,TGT | S,C 10 | NP_001269464.1 | |
NM_013364.5 | 364 | Missense Mutation | AGT,TGT | S,C 10 | NP_037496.4 |