Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCTCAGGCAGAGTGTACTTCTTC[C/T]TGTGCGCACCTTAATAGCAGCCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYTL5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SYTL5 - synaptotagmin like 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163334.1 | Intron | NP_001156806.1 | ||||
NM_001163335.1 | Intron | NP_001156807.1 | ||||
NM_138780.2 | Intron | NP_620135.1 | ||||
XM_011544001.2 | Intron | XP_011542303.1 | ||||
XM_011544002.2 | Intron | XP_011542304.1 | ||||
XM_017029972.1 | Intron | XP_016885461.1 | ||||
XM_017029973.1 | Intron | XP_016885462.1 | ||||
XM_017029974.1 | Intron | XP_016885463.1 |