Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGCTACTGGACTCGCCTCGAATA[C/T]ACTCATCTGTTTGCTTCAAAAGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300267 MIM: 300386 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGEF6 - Rac/Cdc42 guanine nucleotide exchange factor 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306177.1 | 3155 | Missense Mutation | TAT,TGT | Y,C 609 | NP_001293106.1 | |
NM_004840.2 | 3155 | Missense Mutation | TAT,TGT | Y,C 763 | NP_004831.1 | |
XM_005262499.2 | 3155 | Missense Mutation | TAT,TGT | Y,C 714 | XP_005262556.1 | |
XM_011531412.2 | 3155 | Missense Mutation | TAT,TGT | Y,C 790 | XP_011529714.1 | |
XM_011531413.1 | 3155 | Missense Mutation | TAT,TGT | Y,C 767 | XP_011529715.1 | |
XM_011531414.1 | 3155 | Missense Mutation | TAT,TGT | Y,C 760 | XP_011529716.1 | |
XM_011531415.2 | 3155 | Missense Mutation | TAT,TGT | Y,C 733 | XP_011529717.1 | |
XM_011531416.2 | 3155 | Missense Mutation | TAT,TGT | Y,C 636 | XP_011529718.1 | |
XM_011531417.1 | 3155 | Missense Mutation | TAT,TGT | Y,C 613 | XP_011529719.1 | |
XM_017029975.1 | 3155 | Missense Mutation | TAT,TGT | Y,C 794 | XP_016885464.1 | |
XM_017029976.1 | 3155 | Missense Mutation | TAT,TGT | Y,C 560 | XP_016885465.1 |
CD40LG - CD40 ligand | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |