Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAACGCGTACCGCACACCAACGTT[C/T]CCACAGTTTCGGACGCAGTATATCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300512 | ||||||||||||||||||||
Literature Links: |
WDR13 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WDR13 - WD repeat domain 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166426.1 | 227 | UTR 5 | NP_001159898.1 | |||
NM_017883.4 | 227 | Silent Mutation | TTC,TTT | F,F 23 | NP_060353.2 | |
XM_006724539.1 | 227 | Silent Mutation | TTC,TTT | F,F 23 | XP_006724602.1 | |
XM_006724541.3 | 227 | Intron | XP_006724604.1 | |||
XM_011543947.2 | 227 | Silent Mutation | TTC,TTT | F,F 23 | XP_011542249.1 |