Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCGAATTGCCCTGGTCGTGTTTGT[C/T]GGTGGCACCCTAGTTCTGGGCACGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300550 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHEX PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PHEX - phosphate regulating endopeptidase homolog, X-linked | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000444.5 | 643 | Silent Mutation | GTC,GTT | V,V 27 | NP_000435.3 | |
NM_001282754.1 | 643 | Silent Mutation | GTC,GTT | V,V 27 | NP_001269683.1 | |
XM_011545533.1 | 643 | Intron | XP_011543835.1 | |||
XM_011545536.2 | 643 | Intron | XP_011543838.1 | |||
XM_017029579.1 | 643 | Intron | XP_016885068.1 |