Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAGATTGTTGGTTCAAAGGTTAT[C/T]CTAAAGAAAATATATCCATTTGGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300121 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCX PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCX - doublecortin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000555.3 | 3643 | UTR 3 | NP_000546.2 | |||
NM_001195553.1 | 3643 | UTR 3 | NP_001182482.1 | |||
NM_178151.2 | 3643 | UTR 3 | NP_835364.1 | |||
NM_178152.2 | 3643 | UTR 3 | NP_835365.1 | |||
NM_178153.2 | 3643 | UTR 3 | NP_835366.1 | |||
XM_011530878.2 | 3643 | UTR 3 | XP_011529180.1 | |||
XM_011530879.2 | 3643 | UTR 3 | XP_011529181.1 | |||
XM_011530880.2 | 3643 | UTR 3 | XP_011529182.1 | |||
XM_017029312.1 | 3643 | UTR 3 | XP_016884801.1 |