Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGATGTTGAGCTCGGGCTTGCTGT[C/T]GCCATTCTGCTGCTCCCGCAGGCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300089 MIM: 300634 MIM: 300090 | ||||||||||||||||||||
Literature Links: |
IDH3G PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IDH3G - isocitrate dehydrogenase 3 (NAD(+)) gamma | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDZD4 - PDZ domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303512.1 | 2424 | Missense Mutation | AAC,GAC | N,D 719 | NP_001290441.1 | |
NM_001303513.1 | 2424 | Missense Mutation | AAC,GAC | N,D 617 | NP_001290442.1 | |
NM_001303514.1 | 2424 | Missense Mutation | AAC,GAC | N,D 604 | NP_001290443.1 | |
NM_001303515.1 | 2424 | Missense Mutation | AAC,GAC | N,D 638 | NP_001290444.1 | |
NM_001303516.1 | 2424 | Missense Mutation | AAC,GAC | N,D 632 | NP_001290445.1 | |
NM_032512.3 | 2424 | Missense Mutation | AAC,GAC | N,D 713 | NP_115901.2 |
SSR4 - signal sequence receptor subunit 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |