Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGTTTTACTGTATTTGTTCTGTG[C/T]GATGACCTTTCACTCACCTCTGTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300212 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RGN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RGN - regucalcin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282848.1 | Intron | NP_001269777.1 | ||||
NM_001282849.1 | Intron | NP_001269778.1 | ||||
NM_004683.5 | Intron | NP_004674.1 | ||||
NM_152869.3 | Intron | NP_690608.1 | ||||
XM_006724567.2 | Intron | XP_006724630.1 | ||||
XM_006724568.2 | Intron | XP_006724631.1 | ||||
XM_017029954.1 | Intron | XP_016885443.1 |