Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATATCATGCAAAAATAAGCAAAC[C/T]TGAGCAATCTATGGCATCCAGATTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300133 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
VBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VBP1 - VHL binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303543.1 | Intron | NP_001290472.1 | ||||
NM_001303544.1 | Intron | NP_001290473.1 | ||||
NM_001303545.1 | Intron | NP_001290474.1 | ||||
NM_003372.6 | Intron | NP_003363.1 |