Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGACAACTGGGTTTTGGAAGCT[C/G]TGGCGACAGATGAAACACTTGAATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300078 MIM: 300951 | ||||||||||||||||||||
Literature Links: |
NDUFA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NDUFA1 - NADH:ubiquinone oxidoreductase subunit A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004541.3 | 1016 | Intron | NP_004532.1 |
RNF113A - ring finger protein 113A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006978.2 | 1016 | Missense Mutation | CAC,CAG | H,Q 267 | NP_008909.1 |