Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATGGCACTTTACTGAGGAGTTCA[A/G]ATTCCTCAGGTGCCTTTTGAAGTAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300777 | ||||||||||||||||||||
Literature Links: |
TMLHE PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMLHE - trimethyllysine hydroxylase, epsilon | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184797.1 | 1037 | Missense Mutation | TCT,TTT | S,F 289 | NP_001171726.1 | |
NM_018196.3 | 1037 | Missense Mutation | TCT,TTT | S,F 289 | NP_060666.1 | |
XM_011531182.2 | 1037 | Missense Mutation | TCT,TTT | S,F 238 | XP_011529484.1 | |
XM_017029620.1 | 1037 | Intron | XP_016885109.1 |