Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCTGCTGCTCCTTCTGCAGCTTGC[A/G]GATATAATCCACAGAGGCCTTCAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 314310 | ||||||||||||||||||||
Literature Links: |
TFE3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TFE3 - transcription factor binding to IGHM enhancer 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282142.1 | 1480 | Missense Mutation | CGC,TGC | R,C 292 | NP_001269071.1 | |
NM_006521.5 | 1480 | Missense Mutation | CGC,TGC | R,C 397 | NP_006512.2 |