Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCTCGGGCACTTGTGGGGATTC[A/G]GGAGCTGGCTCTTCAGGGCTCAGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300506 | ||||||||||||||||||||
Literature Links: |
TSC22D3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TSC22D3 - TSC22 domain family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001015881.1 | 961 | Silent Mutation | CCC,CCT | P,P 63 | NP_001015881.1 | |
NM_001318468.1 | 961 | Silent Mutation | CCC,CCT | P,P 186 | NP_001305397.1 | |
NM_001318470.1 | 961 | Silent Mutation | CCC,CCT | P,P 186 | NP_001305399.1 | |
NM_004089.3 | 961 | Silent Mutation | CCC,CCT | P,P 120 | NP_004080.2 | |
NM_198057.2 | 961 | Silent Mutation | CCC,CCT | P,P 186 | NP_932174.1 | |
XM_005262099.1 | 961 | Silent Mutation | CCC,CCT | P,P 186 | XP_005262156.1 | |
XM_005262100.1 | 961 | Silent Mutation | CCC,CCT | P,P 186 | XP_005262157.1 | |
XM_005262102.1 | 961 | Silent Mutation | CCC,CCT | P,P 186 | XP_005262159.1 | |
XM_005262103.3 | 961 | Silent Mutation | CCC,CCT | P,P 186 | XP_005262160.1 | |
XM_011530884.1 | 961 | Silent Mutation | CCC,CCT | P,P 186 | XP_011529186.1 | |
XM_017029335.1 | 961 | Silent Mutation | CCC,CCT | P,P 186 | XP_016884824.1 |