Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTCATCCAAAATCTTGCAGTCCC[C/T]GCAGGCCTCCCTCTTAATGAGACAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300003 | ||||||||||||||||||||
Literature Links: |
ARSF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARSF - arylsulfatase F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201538.1 | 469 | Silent Mutation | CCC,CCT | P,P 111 | NP_001188467.1 | |
NM_001201539.1 | 469 | Silent Mutation | CCC,CCT | P,P 111 | NP_001188468.1 | |
NM_004042.4 | 469 | Intron | NP_004033.2 | |||
XM_011545522.1 | 469 | Silent Mutation | CCC,CCT | P,P 128 | XP_011543824.1 | |
XM_011545523.1 | 469 | Silent Mutation | CCC,CCT | P,P 128 | XP_011543825.1 | |
XM_011545524.1 | 469 | Silent Mutation | CCC,CCT | P,P 128 | XP_011543826.1 | |
XM_017029527.1 | 469 | Silent Mutation | CCC,CCT | P,P 111 | XP_016885016.1 | |
XM_017029528.1 | 469 | Silent Mutation | CCC,CCT | P,P 128 | XP_016885017.1 |