Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTGTGGCTTCAGGTGTTTGTTACA[C/T]GAGAACAACATATTACGGACTCAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300335 MIM: 300101 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACE2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACE2 - angiotensin I converting enzyme 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021804.2 | Intron | NP_068576.1 | ||||
XM_011545549.1 | Intron | XP_011543851.1 | ||||
XM_011545551.2 | Intron | XP_011543853.1 | ||||
XM_011545552.1 | Intron | XP_011543854.1 |
BMX - BMX non-receptor tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320866.1 | Intron | NP_001307795.1 | ||||
NM_001721.6 | Intron | NP_001712.1 | ||||
NM_203281.2 | Intron | NP_975010.1 | ||||
XM_017029752.1 | Intron | XP_016885241.1 |