Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCATCCAGATGTCGATTGCCTTCA[C/T]GTAGGACACCTACAACATCCAGCGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
GLRA4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GLRA4 - glycine receptor alpha 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024452.2 | 1364 | Missense Mutation | ATG,GTG | M,V 315 | NP_001019623.2 | |
NM_001172285.1 | 1364 | Intron | NP_001165756.1 |
TMEM31 - transmembrane protein 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182541.2 | 1364 | Missense Mutation | CGT,TGT | R,C 57 | NP_872347.2 |