Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCGCATGTCGTAGATCAATAAACA[C/T]TTGAATAAACGAAAGAAGTTCGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300713 MIM: 300295 MIM: 314375 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OTUD5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OTUD5 - OTU deubiquitinase 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIM2 - Pim-2 proto-oncogene, serine/threonine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006875.3 | Intron | NP_006866.2 |
SLC35A2 - solute carrier family 35 member A2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |