Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTGAAACGTTTCCACCTTTCATC[A/G]TGTTTAAAATTTTTCTTCATACTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300753 | ||||||||||||||||||||
Literature Links: |
APOO PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
APOO - apolipoprotein O | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CXorf58 - chromosome X open reading frame 58 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001169574.1 | 444 | Missense Mutation | ATG,GTG | M,V 115 | NP_001163045.1 | |
NM_152761.2 | 444 | Missense Mutation | ATG,GTG | M,V 115 | NP_689974.2 | |
XM_011545473.2 | 444 | Missense Mutation | ATG,GTG | M,V 115 | XP_011543775.1 | |
XM_011545474.2 | 444 | Intron | XP_011543776.1 |