Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGCTCCCAAGGGAAGAAATGTCAT[C/G]ACGAACCAGCCTCTGGACAGTGACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300167 | ||||||||||||||||||||
Literature Links: |
HEPH PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HEPH - hephaestin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130860.3 | 252 | Silent Mutation | ATC,ATG | I,M 50 | NP_001124332.1 | |
NM_001282141.1 | 252 | Silent Mutation | ATC,ATG | I,M 50 | NP_001269070.1 | |
NM_014799.3 | 252 | Intron | NP_055614.1 | |||
NM_138737.4 | 252 | Silent Mutation | ATC,ATG | I,M 101 | NP_620074.3 | |
XM_006724722.1 | 252 | Silent Mutation | ATC,ATG | I,M 50 | XP_006724785.1 | |
XM_011531073.2 | 252 | Silent Mutation | ATC,ATG | I,M 101 | XP_011529375.1 | |
XM_011531074.2 | 252 | Silent Mutation | ATC,ATG | I,M 101 | XP_011529376.1 | |
XM_011531075.2 | 252 | Silent Mutation | ATC,ATG | I,M 101 | XP_011529377.1 | |
XM_017029998.1 | 252 | Silent Mutation | ATC,ATG | I,M 101 | XP_016885487.1 | |
XM_017029999.1 | 252 | Silent Mutation | ATC,ATG | I,M 50 | XP_016885488.1 | |
XM_017030000.1 | 252 | Silent Mutation | ATC,ATG | I,M 50 | XP_016885489.1 | |
XM_017030001.1 | 252 | Missense Mutation | ATC,ATG | I,M 50 | XP_016885490.1 | |
XM_017030002.1 | 252 | Silent Mutation | ATC,ATG | I,M 50 | XP_016885491.1 | |
XM_017030003.1 | 252 | Silent Mutation | ATC,ATG | I,M 50 | XP_016885492.1 |