Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGTAAAATAAAACATGTTGCTAC[A/G]TACTTAGATGTTTTAAAAATTTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300724 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNKSR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNKSR2 - connector enhancer of kinase suppressor of Ras 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168647.1 | Intron | NP_001162118.1 | ||||
NM_001168648.1 | Intron | NP_001162119.1 | ||||
NM_001168649.1 | Intron | NP_001162120.1 | ||||
NM_014927.3 | Intron | NP_055742.2 | ||||
XM_011545471.2 | Intron | XP_011543773.1 | ||||
XM_011545472.2 | Intron | XP_011543774.1 | ||||
XM_017029358.1 | Intron | XP_016884847.1 |