Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGGTTTCTTTCCTGTATGAATTA[A/T]CTGATGCATACTTAGTGTGGCTTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 314995 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF41 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF41 - zinc finger protein 41 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324139.1 | 2982 | Missense Mutation | ATA,TTA | I,L 653 | NP_001311068.1 | |
NM_001324140.1 | 2982 | Missense Mutation | ATA,TTA | I,L 739 | NP_001311069.1 | |
NM_001324141.1 | 2982 | Missense Mutation | ATA,TTA | I,L 653 | NP_001311070.1 | |
NM_001324142.1 | 2982 | Missense Mutation | ATA,TTA | I,L 741 | NP_001311071.1 | |
NM_001324143.1 | 2982 | Missense Mutation | ATA,TTA | I,L 653 | NP_001311072.1 | |
NM_001324144.1 | 2982 | Missense Mutation | ATA,TTA | I,L 739 | NP_001311073.1 | |
NM_001324145.1 | 2982 | Missense Mutation | ATA,TTA | I,L 653 | NP_001311074.1 | |
NM_001324147.1 | 2982 | Missense Mutation | ATA,TTA | I,L 739 | NP_001311076.1 | |
NM_001324148.1 | 2982 | Missense Mutation | ATA,TTA | I,L 741 | NP_001311077.1 | |
NM_001324149.1 | 2982 | Missense Mutation | ATA,TTA | I,L 653 | NP_001311078.1 | |
NM_001324150.1 | 2982 | Missense Mutation | ATA,TTA | I,L 739 | NP_001311079.1 | |
NM_001324151.1 | 2982 | Missense Mutation | ATA,TTA | I,L 749 | NP_001311080.1 | |
NM_001324152.1 | 2982 | Missense Mutation | ATA,TTA | I,L 653 | NP_001311081.1 | |
NM_001324153.1 | 2982 | Missense Mutation | ATA,TTA | I,L 749 | NP_001311082.1 | |
NM_001324154.1 | 2982 | Missense Mutation | ATA,TTA | I,L 773 | NP_001311083.1 | |
NM_001324155.1 | 2982 | Missense Mutation | ATA,TTA | I,L 781 | NP_001311084.1 | |
NM_001324156.1 | 2982 | Missense Mutation | ATA,TTA | I,L 705 | NP_001311085.1 | |
NM_001324157.1 | 2982 | Missense Mutation | ATA,TTA | I,L 703 | NP_001311086.1 | |
NM_007130.3 | 2982 | Missense Mutation | ATA,TTA | I,L 739 | NP_009061.1 | |
NM_153380.3 | 2982 | Missense Mutation | ATA,TTA | I,L 739 | NP_700359.1 | |
XM_006724550.3 | 2982 | Missense Mutation | ATA,TTA | I,L 773 | XP_006724613.1 | |
XM_006724555.3 | 2982 | Missense Mutation | ATA,TTA | I,L 749 | XP_006724618.1 | |
XM_017029810.1 | 2982 | Missense Mutation | ATA,TTA | I,L 773 | XP_016885299.1 | |
XM_017029811.1 | 2982 | Missense Mutation | ATA,TTA | I,L 773 | XP_016885300.1 | |
XM_017029812.1 | 2982 | Missense Mutation | ATA,TTA | I,L 773 | XP_016885301.1 | |
XM_017029813.1 | 2982 | Missense Mutation | ATA,TTA | I,L 773 | XP_016885302.1 | |
XM_017029814.1 | 2982 | Missense Mutation | ATA,TTA | I,L 773 | XP_016885303.1 | |
XM_017029815.1 | 2982 | Missense Mutation | ATA,TTA | I,L 773 | XP_016885304.1 | |
XM_017029816.1 | 2982 | Missense Mutation | ATA,TTA | I,L 749 | XP_016885305.1 | |
XM_017029817.1 | 2982 | Missense Mutation | ATA,TTA | I,L 739 | XP_016885306.1 |