Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCGGGTGACGGTGGTGCCCAAATC[A/G]GAGGAGGAGGAAGAAGAGGAGGAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ATP1B4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATP1B4 - ATPase Na+/K+ transporting family member beta 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142447.2 | 237 | Silent Mutation | TCA,TCG | S,S 51 | NP_001135919.1 | |
NM_012069.4 | 237 | Silent Mutation | TCA,TCG | S,S 51 | NP_036201.1 | |
XM_017029381.1 | 237 | Silent Mutation | TCA,TCG | S,S 51 | XP_016884870.1 |