Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAAGCCCTATTGAATCTTCCTAG[C/T]TTTAGCTGTGAGGCTTAGAGGTAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300086 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LPAR4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LPAR4 - lysophosphatidic acid receptor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278000.1 | Intron | NP_001264929.1 | ||||
NM_005296.2 | Intron | NP_005287.1 | ||||
XM_005262126.3 | Intron | XP_005262183.1 | ||||
XM_006724639.3 | Intron | XP_006724702.1 | ||||
XM_017029437.1 | Intron | XP_016884926.1 | ||||
XM_017029438.1 | Intron | XP_016884927.1 |