Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGATTGCCTGGGTTCAAATCCTGG[C/T]GCTGCCACTTACTGGCCTTGTGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300292 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOXP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FOXP3 - forkhead box P3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP1R3F - protein phosphatase 1 regulatory subunit 3F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184745.1 | 291 | Intron | NP_001171674.1 | |||
NM_033215.4 | 291 | Intron | NP_149992.3 | |||
XM_005272687.3 | 291 | Intron | XP_005272744.1 | |||
XM_017029932.1 | 291 | UTR 5 | XP_016885421.1 | |||
XM_017029933.1 | 291 | UTR 5 | XP_016885422.1 | |||
XM_017029934.1 | 291 | Intron | XP_016885423.1 | |||
XM_017029935.1 | 291 | Intron | XP_016885424.1 |